Learn More
Karl Hecht Pycnometer, unadjusted 100cm³
Brand:  Karl Hecht 42570040
Pycnometer, unadjusted 100cm³
Product Code. 10003811
  • ONLINE EXCLUSIVE PRICE
    59.85€
    47.23€ / Each
    Save 12.62€
    21% Off
Only null left
Estimated Shipment: 22-01-2026