Learn More
Masterflex™ Barbed Y-Connector, PVDF
Combine these simple, economical fittings with flexible tubing for use in low-pressure applications (generally less than 30 psi). Cole-Parmer™ PVDF Barbed Y-Connectors are designed for quick and easy assembly without tools.
Brand:  Masterflex™ 30703-90
18.34 EUR valid until 2026-12-31
BEST PRICE on Promo! Use promo code "28080" to get your promotional price.
Product Code. 11806643
  • ONLINE EXCLUSIVE PRICE
    22.90€
    22.44€ / Pack of 10
    Save 0.46€
    2% Off
View all promotions
In Stock
Edge
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Description
Description
  • Temperature range: –62 to 129°C
  • Excellent Chemical Resistance
  • Sterilize by ethylene oxide or autoclave
Certificates
Promotions
Promotions
Promo
Expires: 2026-12-31
Outlet Corner
Promo Code: 28080
Exclusive offers subject to immediate product availability and whilst stocks last.
Applying Promo and Adding