Learn More
Cole-Parmer™ PVDF Manifold, 10 port
Expand your flow and line capacity with these manifolds. Choose the number of ports and connections to suit your needs.
Brand:  Cole-Parmer™ 31523-57
 View more versions of this product
Product Code. 11715298
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
  • Constructed of durable PVDF.
  • Ideal for applications requiring excellent chemical compatibly.
  • Available in a variety of configurations
Specifications
Specifications
Manifold
9 3/8 in.
-65°F
3/8 in. x 1/4 in.
1/4 in. NPT(F)
1.25 in.
1.25 in.
230°F
PVDF
23.81 cm
-54°C
3/8 in. NPT(F)
3.18 cm
3.18 cm
110 psi
110°C