Learn More
Gilson™ PVDF Connectors
Shop All Gilson Products
Click to view available options
For Use With:
1 to 2mm tubing I.D. to 2 to 3mm
2 to 3mm tubing I.D. to 2 to 3mm
Unit Size:
Pack of 10
Description
- These products are replacement parts for Gilson HPLC systems
Specifications
Specifications
| Material | PVDF |
| For Use With | 1 to 2mm tubing I.D. to 2 to 3mm |
| Packaging Quantity | 10 Set |
| Product Type | Connector |