Learn More
Saint-Gobain Performance Plastics PVDF Clamp Flow Meter
Click to view available options
Includes:
1/8 NPT Clamp
Unit Size:
Each
Description
- Infrared (IR) turbine rotor reflection results in accurate and repeatable flow measurements
- USP Class VI PVDF material can be gamma sterilized up to 50kGy
- Each sensor is pre-calibrated to reach <1% accuracy
- High resolution square wave output
- Performs an easy and fast exchange of the wetted parts
- Small housing with screw gap to mount onto your device or system
- Tubes can be sterilized up to 150°C (302°F)
- Reduces down-time and prevents cross-contamination
Specifications
Specifications
| For Use With (Equipment) | Flowmeter |
| Type | Flow Sensor (Clamp-mount) |
| Includes | 1/8 NPT Clamp |
| Airflow Rate | 0.03 to 2 L/min. |