Learn More
Sanplatec™ Dry Keeper Non-Auto C-Type Portable Desiccator
Product Code. 11847152
Click to view available options
Material:
PVC, ABS
Unit Size:
Each
Product Code. 11847152
Brand: Sanplatec™ 0031
A non-auto type desiccator with a pan for silica gel for use in dehumidificaton and other applications.
  • Shelf plates can be adjusted in 15 steps at 28 mm intervals to match the size of stored items
  • Slim vertical design does not take up much space
  • Body cabinet of PVC, frame of ABS
  • Latch door type
  • White frame, clear transparent body
  • Inside humidity is slightly different depending on the set room temperature and usage conditions
Specifications
Cabinet Style Vertical
Material PVC, ABS
Product Type Desiccator
Door Style Latch
For Use With (Application) Dehumidification
Model C-3W
No. of Shelves 3
Shelf Positions 15 steps at 28 mm intervals
Shelf Type Plate
Width (Metric) 312 mm
Height (Metric) 508 mm
Depth (Metric) 390 mm