Learn More
Buerkle™ PVC Standard Bung
Product Code. 17270644 Shop All Bürkle Products
Change view
Click to view available options
Height (Metric):
30 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
17270644 30 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Attention! Make sure to pay attention to the List of Chemical Resistance!
Product Code. 17270644 Supplier Buerkle™ Supplier No. 97632834
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Attention! Make sure to pay attention to the List of Chemical Resistance!
Laboratory standard bung made of PVC with good chemical resistance against acids and alkalis.
  • PVC, soft, red
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 30 mm
Length (Metric) 28 mm
Width (Metric) 34 mm
Color Red