Learn More
Hecht Karl™ PVC Staining Bridge
Click to view available options
Includes:
Staining Tray
Unit Size:
Each
Description
- Large type
- Extensible
- For dishes and basins of 38cm to 62cm
- Can be fixed at a slanted or horizontal position
- Complete with staining tray and 2 clamps
- All parts made of PVC
Specifications
Specifications
| Includes | Staining Tray |
| Product Type | Staining Bridge |
| Material | PVC |
| Quantity | 1 Pack |
| Size | 380 x 170 x 80 mm |
| Dimensions (L x W x H) | 380 x 170 x 80mm |