Learn More
Kartell™ Plastilab™ General Purpose Input Trays
Product Code. p-8005881
Click to view available options
Dimensions (L x W x H):
303 x 403 x 63 mm
304 x 404 x 64 mm
350 x 300 x 40 mm
Unit Size:
Each
Product Code. 10585421
Brand: Kartell™ 0095200
Only null left
Add to basket
Add to basket
For storing knick-knacks, pipettes or connectors.
  • Five-compartment tray
  • Central compartment used for pipettes
  • For use in desk drawers
  • Made from PVC
Specifications
Capacity (Metric) 7 mL
Dimensions (L x W x H) 350 x 300 x 40 mm
Color White
Height (Metric) 42 mm (Interior), 40 mm (Exterior)
Length (Metric) 300 mm (Interior), 350 mm (Exterior)
Material PVC
Type Tray
Width (Metric) 355 mm (Interior), 300 mm (Exterior)
For Use With 7mL Bijou bottles or specimen tubes
No. per Pack 1/Pk.
Shape Rectangular
Capacity (English) 0.25 oz.