Learn More
BRAND™ Pursept™ AF Liquid Disinfecting Detergent
Product Code. 15222419 Shop All Brand Products
Click to view available options
Volume (Metric):
5 L
Unit Size:
Pack of 3
Product Code. 15222419
Brand: BRAND™ 44855
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Cleaning and disinfection of floors, walls and fittings in one operation. Appropriate for use in the foods sector.
  • Pleasant, fresh fragrance
  • DGHM/VAH-tested
  • RKI-compliant
  • Aerosol-free disinfectant wipes
  • 100 g Pursept™ AF contains: 12.5 g didecyl-dimethyl-ammoniumchloride, 10.4 g alkylpropylenediaminebis-guanidiniumdiacetate, 1.5 g bis(aminopropyl)-laurylamine, 5 - 15% NTA, < 5% non-ionic surfactants, auxiliary agent
  • Clear, green liquid, fresh smell
  • Density approx.: 1 g/mL
  • pH value (1%): approx. 9.5
  • Surface disinfection:
    • Bacteria and fungi with high organic burden according to DGHM/ VAH directives: 0.25% (2.5 g/L)/ 4 hours, 0.5% (5 g/L)/1 hour, 1% (10 g/L)/ 30 minutes, 2% (20 g/L)/ 15 minutes
    • Limited virucidal efficacy* (incl. HIV, HBV, HCV): 1% (10 g/L)/ 15 minutes
    • Inactivation of human rotaviruses: 0.25% (2.5 g/L)/ 2 minutes
    • Effectiveness against TbB: 2% (20 g/L)/ 1 hour
    • (* As recommended by the RKI, Bundesgesundheitsblatt 01/2004)
TRUSTED_SUSTAINABILITY
Specifications
Container Type Can
Description Clear, green liquid detergent, fresh smell
Volume (Metric) 5 L
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • pursept-AF disinfectant
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : guanidine N,N'''-1,3-propanediylbis-N-coco alkyl derivitives, didecyldimethylammonium chloride, 2-pr