Learn More
Brand™ Pursept™ A Xpress Surface Disinfecting Detergents
Product Code. 10394741 Shop All Brand Products
Click to view available options
Volume (Metric):
1 L
5 L
Container Type:
Bottle
Can
Unit Size:
10 L
5 L
Product Code. 10394741
Brand: Brand™ 44835
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Disinfects in just 15 seconds. BRAND™ Pursept™ A Xpress Surface Disinfecting Detergents are effective against bacteria and fungi with little organic burden with mechanics. Limited virucidal efficacy including HIV, HBV, and HCV after 15 seconds.
Disinfects in just 15 seconds. Refillable pump-atomizer without gas. Pleasant, fresh fragrance.
  • DGHM/VAH tested and RKIcompliant, with aerosol-free application (disinfectant wipes).
  •  
Ingredients:
100 g Pursept™ A Xpress contain: 55 g ethanol, 0.03 g n-alkyl-aminopropyl-glycine, auxiliary agents.
Chemical and physical specifications:
  • Clear, colorless liquid,
  • Density: approx. 0.9 g/ml, pH value: approx. 7.5.
Microbiological efficiency of the undiluted working solution:
  • Surface disinfection: Bacteria and fungi with little organic burden with mechanics acc. to DGHM/VAH directives: 15 seconds.
  • Limited virucidal efficacy* (incl. HIV, HBV, HCV): 15 seconds.
  • Effectiveness against TbB: 30 seconds.
* As recommended by the RKI, Bundesgesundheitsblatt 01/2004.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Disinfectant Detergent
Form Liquid
pH 7.5
Appearance Colorless
Certifications/Compliance DGHM/VAH Tested and RKI Compliant
Composition 100g Pursept™ A Xpress Contain: 55g Ethanol, 0.03g n-Alkyl-Aminopropyl-Glycine, Auxiliary Agen
Container Type Can
For Use With (Application) With Aerosol-Free Application
Quantity 3 Pk.
Volume (Metric) 5 L
compliance-icons compliance-icons
Product Identifier
  • pursept A Xpress
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P271-Use only outdoors or in a well-ventilated area.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Ethanol