missing translation for 'onlineSavingsMsg'
Learn More

Purpurin

Product Code. 10164790 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10164790

Brand: Thermo Scientific Acros 211990250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 81-54-9 | C14H8O5 | 256.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-54-9
Molecular Formula C14H8O5
Molecular Weight (g/mol) 256.2
MDL Number MFCD00001203
InChI Key BBNQQADTFFCFGB-UHFFFAOYSA-N
Synonym purpurin, 1,2,4-trihydroxyanthraquinone, purpurine, verantin, hydroxylizaric acid, smoke brown g, 1,2,4-trihydroxy-9,10-anthracenedione, 1,2,4-trihydroxy-9,10-anthraquinone, 1,2,4-trihydroxyanthrachinon, 9,10-anthracenedione, 1,2,4-trihydroxy
PubChem CID 6683
ChEBI CHEBI:8645
IUPAC Name 1,2,4-trihydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=C(C=C3O)O)O

Specifications

Melting Point 253°C to 256°C
Color Brown to Brown-Red
Flash Point 113°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 g
Beilstein 08, 509
Merck Index 15, 8056
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in hot water,freely soluble in alcohol and ether,soluble in benzene,toluene; xylene,boiling alum,solution
Formula Weight 256.2
Physical Form Powder
Chemical Name or Material Purpurin
Show More Show Less
compliance-icons
Product Identifier
  • Purpurin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.