missing translation for 'onlineSavingsMsg'
Learn More

Puromycin Dihydrochloride,, MP Biomedicals

Product Code. 11427620 Shop All MP Biomedical Products
Click to view available options
Quantity:
10 mg
25 mg
100 mg
250 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
250mg
25mg
500mg
This item is not returnable. View return policy

Product Code. 11427620

Brand: MP Biomedicals 0210055283

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 58-58-2
Molecular Formula C22H31Cl2N7O5
Molecular Weight (g/mol) 544.43
MDL Number MFCD00150080
InChI Key MKSVFGKWZLUTTO-MLYJQVMKNA-N
Synonym Stylomycin, 6-dimethylamino-9-(3'-deoxy-3'-(p-methoxy-L-phenylalanyamino)- B-D-ribofuranosyl)-purine
PubChem CID 131632508
SMILES Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1

Specifications

Type Puromycin Dihydrochloride
Physical Form Powder
Extinction Coefficient 20.3 (275nm, 0.1N NaOH), 19.5 (268nm, 0.1 NHCl) (Lit.)
Color White
Quantity 250 mg
IUPAC Name (2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride
Formula Weight 544.4
Chemical Name or Material Puromycin Dihydrochloride, from Streptomyces albo-niger
compliance-icons
Product Identifier
  • Puromycin Dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Puromycin Dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.