Learn More
Puromycin dihydrochloride, 98%, Thermo Scientific™
Puromycin dihydrochloride hydrate, CAS # 58-58-2, is an aminoglycoside showing antibiotic, anti-neoplastic, and anti-protozoal activities. This compound also has the ability to inhibit protein synthesis.
Brand: Thermo Scientific Alfa Aesar J61278.ME
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Information
• Puromycin dihydrochloride hydrate is the hydrochloride salt form of puromycin and is an aminoglycoside with antibiotic activity isolated from the Streptomyces alboniger
• Puromycin dihydrochloride hydrate is a protein synthesis inhibitor that causes premature chain termination by acting as an analog of the 3’-terminal ends of aminoacyl-tRNA, inhibiting protein synthesis
Applications
• This compound shows anti-microbial, anti-protozoal, anti-trypanosomal, and anti-neoplastic activities and is also used as an antibiotic in cell culture
• It also reveals anti-tumor activity in studies on brain cancer cells
• This compound has been used to study transcriptional regulatory mechanisms that control the sequential and coordinate expression of genes during cell differentiation
• It is active against gram-positive microorganisms, less active against acid-fast bacilli and gram-negative microorganisms. Puromycin can prevent growth of bacteria, protozoa, algae, and mammalian cells in in vitro studies.
Safety and Handling
- Puromycin dihydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Puromycin dihydrochloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.