Learn More
Thermo Scientific™ Purified Water for Microbiology
Product Code. 15844917
Change view
Click to view available options
Fill Weight:
10.01 to 0.3 g
100.0g + 1.5g
200.0g + 4.0g
3.0g + 0.3 g
5.0g + 0.3g
Packaging Type:
10 sirop bottles with screw cap closures in a box
10 sirop bottles with screw cap closures in a box.
24 bijou bottles with screw cap closures in a box.
24 plastic universal bottles with screw cap closures in a box.
300 tubes with screw cap closures in a box
Unit Size:
Pack of 10
Pack of 24
Pack of 300
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Packaging Type Unit Size
15844917 10.01 to 0.3 g 24 plastic universal bottles with screw cap closures in a box. Pack of 24
12746919 3.0g + 0.3 g 300 tubes with screw cap closures in a box Pack of 300
13275639 5.0g + 0.3g 24 bijou bottles with screw cap closures in a box. Pack of 24
12756929 100.0g + 1.5g 10 sirop bottles with screw cap closures in a box. Pack of 10
12766929 200.0g + 4.0g 10 sirop bottles with screw cap closures in a box Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15844917 Supplier Thermo Scientific™ Supplier No. BO1336E
  • ONLINE EXCLUSIVE PRICE
    102.00€
    98.99€ / Pack of 24
    Save 3.01€
    2% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Perform sample dilution for microbial examination or ensure sterility of your devices and equipment with Purified Water for Microbiology.
Perform sample dilution for microbial examination or ensure sterility of your devices and equipment with Thermo Scientific™ Oxoid™ Purified Water. The bottles come with screw cap closure for ease of operation.
Use Purified Water as a diluting and rinsing fluid.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Color Colorless, Clear
Content And Storage 2°C to 25°C
Format Plastic universal bottle
Manufacturing Origin United Kingdom
Fill Weight 10.01 to 0.3 g
Packaging Type 24 plastic universal bottles with screw cap closures in a box.
Quantity 24/Pk.