Learn More
RETSCH Pure 70mm Diameter Stainless Steel Ring Sieve For Ultra Centrifugal Mill ZM 200
Shop All Retsch Products
Click to view available options
Pore Size:
0.12 mm
0.2 mm
0.25 mm
0.28 mm
0.5 mm
0.75 mm
1 mm
1.5 mm
2 mm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 2 mm |
| Type | Ring Sieve |
| Material | Stainless Steel |
| For Use With (Equipment) | Ultra Centrifugal Mill ZM 200 |
| Diameter (Metric) Outer | 70 mm |