Learn More
Buerkle™ Pump Station for AccuOne/EnergyOne Pump
Product Code. 11991591 Shop All Bürkle Products
Change view
Click to view available options
Depth (Metric):
50 cm
70 cm
100 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Unit Size
11991591 50 cm Each
11901601 70 cm Each
11941581 100 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11991591 Supplier Buerkle™ Supplier No. 59003050
  • ONLINE EXCLUSIVE PRICE
    194.00€
    184.25€ / Pack of 1
    Save 9.75€
    5% Off
Estimated Shipment: 02-12-2026
To aid in transferring small quantities of watery media up to 300 mPas from canisters, barrels and narrow-necked containers from 32 mm opening diameter.
  • Allows multiple pumping stations with only one pump
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Applications For dispensing acids and bases
Depth (Metric) 50 cm
Product Type Pump Station
For Use With (Equipment) AccuOne/EnergyOne Pump
Quantity 1 Ea.