Learn More
Munktell Filtrak™ Pulp Testing Blotter
Click to view available options
Dimensions (L x W):
170 x 170 mm
Unit Size:
Pack of 200
Description
- Basis Weight: 250 ±25g/m²
- Klemm Absorbency: 70 ±20mm
- Dimensional Change: <3%
- Water Uptake: 450 ±50g/m²
Specifications
Specifications
| Grade | Grade 1600 |
| Dimensions (L x W) | 170 x 170 mm |