missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PTH Antibody Pair [HRP]

Product Code. 18667621 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18667621

Brand: Novus Biologicals™ NBP2794115Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Human PTH.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 312.5 pg/mL
Assay Sensitivity 312.5-20000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human PTH.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1
Gene ID (Entrez) 5741
Gene Symbol PTH
Host Species Rabbit/Mouse
Kit Contents
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human PTH polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4°C).
  • Capture Antibody: 0.2 mg/mL of mouse anti-Human PTH monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 380 ng of recombinant Human PTH.
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Apoptosis, Biologically Active Proteins, Cancer
    Storage Requirements Storage is content dependent.
    Target PTH
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 20000 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.