Learn More
Kartell™ PTFE Watch Glass
Use a watch glass to evaporate liquids, hold solids when weighing a sample, and in situations where cross-contamination could occur. Kartell™ PTFE Watch Glass has good heat and chemical resistance.
Brand:  Kartell™ 0064500
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10254041
  • 45.90€ / Pack of 2
Estimated Shipment: 06-08-2024
Description
  • Superior chemical, mechanical, and heat resistance
  • Can be used safely for continuous service at temperatures up to 250°C
  • Suitable for foodstuff
Specifications
Specifications
Round
Round
99.39 mm
PTFE
Watch Glass