Learn More
VITLAB™ PTFE Watch Glass
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | PTFE |
| Shape | Round |
| Type | Watch Glass |
| For Use With (Application) | Used as Beaker Covers |
| Diameter (Metric) | 50 mm |
Product Title
¿Detecta una oportunidad de mejora?