Learn More
Kartell™ PTFE Supple Connectors
Product Code. 10021061
Change view
Click to view available options
Material:
PTFE
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10021061 PTFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10021061 Supplier Kartell™ Supplier No. 0104800
  • ONLINE EXCLUSIVE PRICE
    133.00€
    116.90€ / Pack of 1
    Save 16.10€
    12% Off
Estimated Shipment: 02-12-2026
Manufactured from PTFE. Kartell™ PTFE Supple Connectors are specially designed to connect to misaligned glass tubes. The connectors are chemically inert and suitable for food contents.
  • PTFE
  • Suitable for food contents
  • Taper conforms to DIN 12242
  • Chemically inert
Specifications
Certifications/Compliance Yes
Material PTFE
For Use With Misaligned glass tubes
Height (Metric) 29.63 mm
Product Type Supple Connector