Learn More
Fisherbrand™ PTFE Stopper, Flask with Handle
Fisherbrand™ PTFE Stopper, Flask with Handle are for use with volumetric flasks and similar containers.
10.08€ - 38.74€
Specifications
Material | PTFE |
---|---|
For Use With (Equipment) | Standard Taper Ground Glass Joints |
Quantity | 1 |
Temperature Range (Metric) | -200°C to +280°C |
Product Code | Brand | Size | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Color | Price | Quantity & Availability | ||||
16255961
|
Fisherbrand™
|
8 (US) | Gray |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
16265961
|
Fisherbrand™
|
9 (US) | Black |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
16275961
|
Fisherbrand™
|
13 (US) | Orange |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
16285961
|
Fisherbrand™
|
16 (US) | White |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
16295961
|
Fisherbrand™
|
19 (US) | Blue |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
16205971
|
Fisherbrand™
|
22 (US) | Green |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
16215971
|
Fisherbrand™
|
27 (US) | Yellow |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
16225971
|
Fisherbrand™
|
32 (US) | Red |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
16235971
|
Fisherbrand™
|
38 (US) | Black |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
- Solid Construction
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifications
PTFE | |
Standard Taper Ground Glass Joints | |
1 | |
-200°C to +280°C |