Learn More
Fisherbrand™ PTFE Stopper, Flask with Handle
Fisherbrand™ PTFE Stopper, Flask with Handle are for use with volumetric flasks and similar containers.
Brand:  Fisherbrand™
Additional Details : Weight : 0.03500kg
 View more versions of this product
Product Code. 16225971
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • Solid Construction
  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications
Specifications
Fisherbrand™ PTFE Stopper, Flask with Handle, Red Handle
32 (US)
PTFE
Standard Taper Ground Glass Joints
Red
1
-200°C to +280°C