Learn More
Bohlender™ PTFE Stirrer Shafts with Blade
Click to view available options
Length (Metric):
800 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Shaft | 10 mm |
| For Use With (Application) | Tangential flow with little turbulence, gentle stirring |
| Includes | with Blade |
| Length (Metric) | 800 mm |
| Quantity | 1 Each |
| Type | Stirring Shaft |
| Material | PTFE |