Learn More
RETSCH PTFE with Steel Core Grinding Balls
Product Code. 10339540 Shop All Retsch Products
Change view
Click to view available options
Diameter (Metric):
0.9 mm
12 mm
20 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10339540 12 mm Each
10257990 0.9 mm Each
10572111 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10339540 Supplier RETSCH Supplier No. 05.368.0046
Specifications
Product Type Grinding Balls
Diameter (Metric) 12 mm
For Use With (Equipment) MM 200 and MM 301
Material Teflon with Steel Core