Learn More
Wheaton™ PTFE/Silicone Septa
Shop All Wheaton Science Products Products
Click to view available options
Diameter (Metric) Outer:
8 mm
13 mm
15 mm
18 mm
20 mm
Unit Size:
Pack of 100
Pack of 120
Specifications
Specifications
| Material (Liner) | PTFE/Silicone |
| Color | Natural |
| Diameter (Metric) Outer | 8 mm |
| Material | PTFE, Silicone |
| Autoclavable | Autoclavable |
| Quantity | 120 Pk. |
| Type | Septa |