Learn More
BRAND™ PTFE Seal
Product Code. p-8004644 Shop All Brand Products
Click to view available options
Volume (Metric):
100 and 200 μL
20 and 25 μL
50 μL
Unit Size:
Pack of 3
Product Code. 10568331
Brand: BRAND™ 701924
Only null left
Add to basket
Add to basket
For use with the Transferpettor™ line, specially designed to handle viscous, foaming liquids and high vapor pressure.
  • With DE-M marking
  • Provided with mounting block
  • Provided in packs of 3
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Conformity certified
Material PTFE
Volume (Metric) 100 and 200 μL
For Use With (Equipment) For Transferpettor™ 100, 200μL capacity
Includes Mounting block
Quantity 3/Pk.