Learn More
DWK Life Sciences PTFE Hose Connection, with silicone seal (VMQ), complete with screw cap PBT, straight
For safe connection of hoses.
Brand:  DWK Life Sciences 286230005
69.11 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 10376272
  • 72.75€ / Pack of 10
Estimated Shipment: 05-06-2024
Description
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PTFE for use with higher thermal and chemical resistance requirements.
  • Hose olives enable the connection of hoses to laboratory apparatuses
Specifications
Specifications
10/Pk.
Straight
9.0 mm
GL 14