Learn More
Horst™ PTFE HBSI Model Hard-Wearing Heating Tape
Click to view available options
Length (Metric):
0.8 m
1.5 m
2 m
3 m
4 m
Unit Size:
Each
Specifications
Specifications
| Dimensions (L x W) | 0.8 Lm |
| Dimensions (L x W x H) | 0.8 Lm |
| Length (Metric) | 0.8 m |
| Electrical Requirements | ∼230V Voltage |
| Model | HBSI |
| Product Type | Hard-wearing Heating Tape |