Learn More
Kartell™ Lining for Glass Tapers
Product Code. 10554851
Click to view available options
Tubing Size:
14/23 Cone Size
19/26 Cone Size
24/29 Cone Size
29/32 Cone Size
45/40 Cone Size
Unit Size:
Pack of 10
Product Code. 10554851
Brand: Kartell™ 0103800
Constructed of PTFE for high temperature and chemical resistance. Kartell™ Lining for Glass Tapers are specially designed to prevent machined glass joints from binding.
  • Designed to prevent machined glass joints from binding
  • Conforms to DIN 12242
  • Suitable for foodstuff
Specifications
Certifications/Compliance DIN 12242
Material PTFE
Diameter (Metric) Outer 12 mm
For Use With Glass Joint
For Use With (Application) Prevent glass joints from binding
Packaging Quantity 10/Pk.
Tubing Size 14/23 Cone Size
Product Type Ground Joint Sleeve Lining