missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ PTFE Cylindrical Magnetic Stirring Bar

Use this magnetic stirring bar for stirring and mixing in liquid media, over a broad temperature range and with virtually any chemical agent. Bohlender™ BOLA™ PTFE Cylindrical Magnetic Stirring Bars offer excellent centering and smooth running characteristics.

Brand:  Bohlender™ C350-21

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10119730

  • 25.20€ / Pack of 10

Estimated Shipment: 15-11-2023
to see stock.
Add to basket

Description

Description

  • Cylindrical magnetic stirring bars
  • PTFE-encapsulated magnetic core (Alnico 5)
  • Standard magnetic stirring bar
  • Use in open and closed systems and under pressure or vacuum
  • Temperature resistance: -200 to +250°C
  • Universal chemical resistance
Specifications

Specifications

Alnico 5 magnetic core
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics
PTFE (Polytetrafluoroethylene)
30mm
6mm
1/Pk.
FDA Certified
Cylindrical
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers