Learn More
PTFE Beaker;Cap (ml), 250;Dia (mm), 66;Ht (mm), 97
Product Code. p-6931024
Click to view available options
For Use With (Equipment):
1000mL beaker
100mL beaker
10mL beaker
2000mL beaker
250mL beaker
25mL beaker
500mL beaker
50mL beaker
Unit Size:
Each
Product Code. 15461477
Brand: Thermo Scientific Alfa Aesar 038084.KT
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with beakers
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Available in a range of diameters
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer 24mm
Material PTFE
For Use With (Equipment) 10mL beaker
Height (Metric) 33mm