Learn More
Aalborg™ PTFE Aluminum Shell Needle Valve
Description
- Fluids in contact with PTFE only
- Structurally rigid metal shell.
- One PTFE o-ring
- Simplicity - only seven components.
- Up to 55 SLPM (air) or 2.8 LPM (water)
Specifications
Specifications
| Type | Needle Valve |
| Includes | Glass Nipple Fitting |
| Airflow Rate | 55 L/min. |
| Waterflow Rate | 28 mL/min. |
| Max. Operating Temperature (Metric) | 65°C |