Learn More
Kinematica Dispersing Aggregate Dia. 30mm PT-DA 30/2WEC-F250
Brand:  Kinematica 11030309
Dispersing Aggregate Dia. 30mm PT-DA 30/2WEC-F250
Product Code. 15503940
  • ONLINE EXCLUSIVE PRICE
    3433.50€
    3189.00€ / Each
    Save 244.50€
    7% Off
Only null left
Estimated Shipment: 04-02-2026