missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Temperature Measurement of the Specimen
Shop All BINDER Products
Click to view available options
Material:
Copper
Unit Size:
Each
Description
- Sensor with steel-covered, heat-resistant line
- Additional indication of the specimen temperature on the display
Specifications
Specifications
| Description | Temperature measurement of the specimen - with flexible Pt 100 sensor and digital specimen temperature display |
| For Use With (Equipment) | BINDER Chambers Series VD, VDL |
| Material | Copper |
| Type | Sensor |
| Includes | Flexible Pt 100 sensor and digital specimen temperature display |
| Weight (English) | 20.94 lb. |
| Weight (Metric) | 9.5 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction