Learn More
PSPC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA597193
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: MMLRGNLKQV RIEKNPARLR ALESAVGESE PAAAAAMALA LAGEPAPPAP APPEDHPDEE MGFTIDIKSF; Positive Samples: BxPC-3, A-549, U-87MG, Mouse brain; Cellular Location: Cytoplasm, Nucleus, Nucleus matrix, Nucleus speckle, nucleolus
Regulates, cooperatively with NONO and SFPQ, androgen receptor-mediated gene transcription activity in Sertoli cell line. Binds to poly(A), poly(G) and poly(U) RNA homopolymers.Specifications
PSPC1 | |
Polyclonal | |
Unconjugated | |
PSPC1 | |
PSPC1 | |
Recombit fusion protein containing a sequence corresponding to amino acids 1-70 of human PSPC1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
0.68 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R326, Q8WXF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55269, 66645 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |