Learn More
PSPC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558585
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PAMGPEGAAN MGTPMMPDNG AVHNDRFPQG PPSQMGSPMG SRTGSETPQA PMSGVGPVSG GPGGFGRGSQ GGNFEGPNKR RRY Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 96%.
Regulates, cooperatively with NONO and SFPQ, androgen receptor-mediated gene transcription activity in Sertoli cell line. Binds to poly(A), poly(G) and poly(U) RNA homopolymers.Specifications
PSPC1 | |
Polyclonal | |
Unconjugated | |
PSPC1 | |
5730470C09Rik; AI327109; AI449052; mPSP1; Paraspeckle component 1; paraspeckle protein 1; PSP1; Pspc1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
305910, 55269, 66645 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q4KLH4, Q8R326, Q8WXF1 | |
PSPC1 | |
Recombinant protein corresponding to Human PSPC1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only