missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PSMD3 (Human) Recombinant Protein

Human PSMD3 full-length ORF ( AAH20518.1, 1 a.a. - 534 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005709-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193645

  • 528.00€ / 25µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

  • Sequence: MKQEGSARRRGADKAKPPPGGGEQEPPPPPAPQDVEMKEEAATGGGSTGEADGKTAAAAAEHSQRELDTVTLEDIKEHVKQLEKAVSGKEPRFVLRALRMLPSTSRRLNHYVLYKAVQGFFTSNNATRDFLLPFLEEPMDTEADLQFRPRTGKAASTPLLPEVEAYLQLLVVIFMMNSKRYKEAQKISDDLMQKISTQNRRALDLVAAKCYYYHARVYEFLDKLDVVRSFLHARLRTATLRHDADGQATLLNLLLRNYLHYSLYDQAEKLVSKSVFPEQANNNEWARYLYYTGRIKAIQLEYSEARRTMTNALRKAPQHTAVGFKQTVHKLLIVVELLLGEIPDRLQFRQPSLKRSLMPYFLLTQAVRTGNLAKFNQVLDQFGEKFQADGTYTLIIRLRHNVIKTGVRMISLSYSRISLADIAQKLQLDSPEDAEFIVAKAIRDGVIEASINHEKGYVQSKEMIDIYSTREPQLAFHQRISFCLDIHNMSVKAMRFPPKSYNKDLESAEERREREQQDLEFAKEMAEDDDDSFP
Specifications

Specifications

AAH20518.1
proteasome (prosome, macropain) 26S subunit, non-ATPase, 3
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PSMD3
GST
5709
Wheat germ expression system
25 μg
P58, RPN3, S3
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers