missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PSMD2 (Human) Recombinant Protein

Human PSMD2 full-length ORF ( AAH02368, 1 a.a. - 908 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005708-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16119821

  • 545.00€ / 25µg

Estimated Shipment: 04-10-2022
to see stock.
Add to basket

Description

Description

  • Sequence: MEEGGRDKAPVQPQQSPAAAPGGTDEKPSGKERRDAGDKDKEQELSEEDKQLQDELEMLVERLGEKDTSLYRPALEELRRQIRSSTTSMTSVPKPLKFLRPHYGKLKEIYENMAPGENKRFAADIISVLAMTMSGERECLKYRLVGSQEELASWGHEYVRHLAGEVAKEWQELDDAEKVQREPLLTLVKEIVPYNMAHNAEHEACDLLMEIEQVDMLEKDIDENAYAKVCLYLTSCVNYVPEPENSALLRCALGVFRKFSRFPEALRLALMLNDMELVEDIFTSCKDVVVQKQMAFMLGRHGVFLELSEDVEEYEDLTEIMSNVQLNSNFLALARELDIMEPKVPDDIYKTHLENNRFGGSGSQVDSARMNLASSFVNGFVNAAFGQDKLLTDDGNKWLYKNKDHGMLSAAASLGMILLWDVDGGLTQIDKYLYSSEDYIKSGALLACGIVNSGVRNECDPALALLSDYVLHNSNTMRLGSIFGLGLAYAGSNREDVLTLLLPVMGDSKSSMEVAGVTALACGMIAVGSCNGDVTSTILQTIMEKSETELKDTYARWLPLGLGLNHLGKGEAIEAILAALEVVSEPFRSFANTLVDVCAYAGSGNVLKVQQLLHICSEHFDSKEKEEDKDKKEKKDKDKKEAPADMGAHQGVAVLGIALIAMGEEIGAEMALRTFGHLLRYGEPTLRRAVPLALALISVSNPRLNILDTLSKFSHDADPEVSYNSIFAMGMVGSGTNNARLAAMLRQLAQYHAKDPNNLFMVRLAQGLTHLGKGTLTLCPYHSDRQLMSQVAVAGLLTVLVSFLDVRNIILGKSHYVLYGLVAAMQPRMLVTFDEELRPLPVSVRVGQAVDVVGQAGKPKTITGFQTHTTPVLLAHGERAELATEEFLPVTPILEGFVILRKNPNYDL
Specifications

Specifications

AAH02368
proteasome (prosome, macropain) 26S subunit, non-ATPase, 2
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PSMD2
GST
5708
Wheat germ expression system
25 μg
MGC14274, P97, Rpn1, S2, TRAP2
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers