missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMB9 (Human) Recombinant Protein
Human PSMB9 full-length ORF ( AAH65513, 1 a.a. - 219 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005698-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MLRAGAPTGDLPRAGEVHTGTTIMAVEFDGGVVMGSDSRVSAGEAVVNRVFDKLSPLHERIYCALSGSAADAQAVADMAAYQLELHGIELEEPPLVLAAANVVRNISYKYREDLSAHLMVAGWDQREGGQVYGTLGGMLTRQPFAIGGSGSTFIYGYVDAAYKPGMSPEECRRFTTDAIALAMSRDGSSGGVIYLVTITAAGVDHRVILGNELPKFYDE
Specifications
AAH65513 | |
proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctional peptidase 2) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMB9 | |
GST |
5698 | |
Wheat germ expression system | |
25 μg | |
LMP2, MGC70470, PSMB6i, RING12, beta1i | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |