missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMA7 (Human) Recombinant Protein
Human PSMA7 full-length ORF ( AAH04427, 1 a.a. - 248 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005688-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSYDRAITVFSPDGHLFQVEYAQEAVKKGSTAVGVRGRDIVVLGVEKKSVAKLQDERTVRKICALDDNVCMAFAGLTADARIVINRARVECQSHRLTVEDPVTVEYITRYIASLKQRYTQSNGRRPFGISALIVGFDFDGTPRLYQTDPSGTYHAWKANAIGRGAKSVREFLEKNYTDEAIETDDLTIKLVIKALLEVVQSGGKNIELAVMRRDQSLKILNPEEIEKYVAEIEKEKEENEKKKQKKAS
Specifications
AAH04427 | |
proteasome (prosome, macropain) subunit, alpha type, 7 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMA7 | |
GST |
5688 | |
Wheat germ expression system | |
10 μg | |
C6, HSPC, MGC3755, RC6-1, XAPC7 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |