missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMA3 (Human) Recombinant Protein
Human PSMA3 full-length ORF ( AAH05265, 1 a.a. - 248 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005684-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSSIGTGYDLSASTFSPDGRVFQVEYAMKAVENSSTAIGIRCKDGVVFGVEKLVLSKLYEEGSNKRLFNVDRHVGMAVAGLLADARSLADIAREEASNFRSNFGYNIPLKHLADRVAMYVHAYTLYSAVRPFGCSVNDGAQLYMIDPSGVSYGYWGCAIGKARQAAKTEIEKLQMKEMTCRDIVKEVAKIIYIVHDEVKDKAFELELSWVGELTNGRHEIVPKDIREEAEKYAKESLKEEDESDDDNM
Specifications
AAH05265 | |
proteasome (prosome, macropain) subunit, alpha type, 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMA3 | |
GST |
5684 | |
Wheat germ expression system | |
25 μg | |
HC8, MGC12306, MGC32631, PSC3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |