Specifications
| For Use With (Application) | Immunocytochemistry/Immunofluorescence, In vitro assay, Live Imaging Microscopy |
| Product Type | Polarity Sensitive Indicator of Viability Apoptosis Microscopy Kit |
| Content And Storage | Propidium Iodide Staining Solution (Full Size: 500μL / Sample Size: 125μL), pSIVA-IANBD (Full Size: 200μL / Sample Size: 50μL). 4°C |
| Conjugate | IANBD |
| Quantity | 1 Kit |