Learn More
Pseudo Erythromycin A Enol Ether, TRC
Product Code. 30133282
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133282 1 mg 1mg
17928424 25 mg 25mg
17929264 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133282 Supplier Toronto Research Chemicals Supplier No. TRCP8395001MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 105882-69-7
Molecular Formula C37 H65 N O12
Molecular Weight (g/mol) 715.91
Synonym 4,13-Dioxabicyclo[8.2.1]tridec-12-en-5-one, 7-[(2,6-dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-3-[(1R,2R)-1,2-dihydroxy-1-methylbutyl]-2,6,8,10,12-pentamethyl-9-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-, (2R,3R,6R,7S,8S,9R,10R)-, 4,13-Dioxabicyclo[8.2.1]tridec-12-en-5-one, 7-[(2,6-dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-3-(1,2-dihydroxy-1-methylbutyl)-2,6,8,10,12-pentamethyl-9-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-, [2R-[2R*,3R*(1R*,2R*),6R*,7S*,8S*,9R*,10R*]]-, (2R,3R,6R,7S,8S,9R,10R)-7-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-3-[(1R,2R)-1,2-dihydroxy-1-methylbutyl]-2,6,8,10,12-pentamethyl-9-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-4,13-dioxabicyclo[8.2.1]tridec-12-en-5-one, 8,9-Anhydropseudo-erythromycin A-6,9-hemiketal, EM 701, LY 267108, Pseudoerythromycin A enol ether
IUPAC Name (2R,3R,6R,7S,8S,9R,10R)-3-[(2R,3R)-2,3-dihydroxypentan-2-yl]-9-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-7-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-2,6,8,10,12-pentamethyl-4,13-dioxabicyclo[8.2.1]tridec-1(12)-en-5-one
SMILES CC[C@@H](O)[C@@](C)(O)[C@@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@]4(C)CC(=C(O4)[C@@H]1C)C
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C37H65NO12/c1-14-25(39)37(10,43)32-20(4)28-18(2)16-36(9,50-28)31(49-34-27(40)24(38(11)12)15-19(3)45-34)21(5)29(22(6)33(42)48-32)47-26-17-35(8,44-13)30(41)23(7)46-26/h19-27,29-32,34,39-41,43H,14-17H2,1-13H3/t19-,20+,21+,22-,23+,24+,25-,26+,27-,29+,30+,31-,32-,34+,35-,36-,37-/m1/s1
Formula Weight 715.4507
Percent Purity >95%
Chemical Name or Material Pseudo Erythromycin A Enol Ether
Grade HPLC