Learn More
Prulifloxacin, TRC
Product Code. 30415003
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415003 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30415003 Supplier Toronto Research Chemicals Supplier No. TRCP83888550MG
 Restricted Item
Estimated Shipment: 02-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Prulifloxacin
CAS 123447-62-1
Purity Grade Notes HPLC
Molecular Formula C21 H20 F N3 O6 S
Quantity 50 mg
Synonym 1H,4H-[1,3]Thiazeto[3,2-a]quinoline-3-carboxylic acid','6-fluoro-1-methyl-7-[4-[(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl]-1-piperazinyl]-4-oxo-, 1,3-Dioxole','1H,4H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid deriv., 6-Fluoro-1-methyl-7-[4-[(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl]-1-piperazinyl]-4-oxo-1H,4H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid, (±)-7-[4-[(Z)-2,3-Dihydroxy-2-butenyl]-1-piperazinyl]-6-fluoro-1-methyl-4-oxo-1H,4H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid','cyclic carbonate, NM 441, Prulifloxacin, Quisnon, Sword
InChI Formula InChI=1S/C21H20FN3O6S/c1-10-16(31-21(29)30-10)9-23-3-5-24(6-4-23)15-8-14-12(7-13(15)22)18(26)17(20(27)28)19-25(14)11(2)32-19/h7-8,11H,3-6,9H2,1-2H3,(H,27,28)
SMILES CC1SC2=C(C(=O)O)C(=O)c3cc(F)c(cc3N12)N4CCN(CC5=C(C)OC(=O)O5)CC4
IUPAC Name 6-fluoro-1-methyl-7-[4-[(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl]piperazin-1-yl]-4-oxo-1H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid
Molecular Weight (g/mol) 461.46
Formula Weight 461.1057
Percent Purity >95