Learn More
Hamilton™ PRP-1 Reversed-Phase HPLC Column, 100 x 250 mm, 12-20 μm
Click to view available options
Length (Metric):
250 mm
Unit Size:
Each
Description
Specification Sheet
- Separation Mode – Reversed-Phase
- Stationary Phase – PRP-1
- Format – HPLC Column
- Hardware Inner Diameter – 100 mm
- Hardware Length – 250 mm
- Particle Size – 12-20 μm
- Packing Material Type – PSDVB
- Pore Size – 100 Å
- Hardware Material – Stainless Steel
- Max Pressure – 2,000 psi
- Mobile Phase Limits – pH 1-13. 0-100% aqueous, organic modifier
- Buffer Strength – 0.0-0.5N
- Temp Limits – 5-85°C
- Restoration – Run a gradient of 100% water to 100% acetonitrile. Repeat three times
- Compound Classification – Organics
- UOM – 1 EACH
Specifications
Specifications
| Column Format | HPLC Column |
| Diameter (Metric) | 100 mm |
| Length (Metric) | 250 mm |
| Max. Pressure | 2,000 psi |
| Particle Size | 12 to 20 μm |
| pH | pH 1-13. 0-100% aqueous, organic modifier |
| Pore Size | 100 Å |
| Temperature | 5° to 85°C |
| Packing Material | PSDVB |
| Column Type | PRP-1 Columns |
| Show More |