Learn More
Macherey-Nagel™ Protino Ni-TED 150 kit for His-tag Protein Purification in Mini format
Product Code. 10316642
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 10
Pack of 50
Product Code. 10316642
Brand: Macherey-Nagel™ 745100.10
The matrix of choice for highest protein purity in Mini format.
Protino™ NI-TED, the purer alternative for His-tag protein purification
  • Chelating group TED, one protein binding site
  • High stability against reducing/chelating agents
  • Low metal leaching
  • High protein purity
  • High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
  • Dry resin, storage at room temperature
Protino™ NI-TED packed columns
  • Ready to use pre-packed columns filled with dry Protino™ Ni-TED resin
  • Buffers included
Protino™ NI-TED resin
  • Dry bulk matrix
  • Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink well - highest flexibility
  • Batch procedure, gravity flow column chromatography, FPLC™ applications
  • Empty Protino™ columns available for use with Protino™ Ni-TED resin
  • Empty 96 well receiver plates available for use with Protino™ Ni-TED resin
For purification of polyhistidine-tagged proteins; batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.
Specifications
Bed Volume 80 μL
Binding Capacity 400μg
Kit Contents Protino Ni-TED 150 Packed Columns, buffers
Matrix Macroporous silica
Content And Storage Room Temperature
Quantity 1 Kit
For Use With (Application) Purification of polyhistidine-tagged proteins
Type Ni-TED Protein Purification Kit
Ligand Type TED (tris-carboxymethyl ethylene diamine)
Sufficient For 10 preps
compliance-icons
Product Identifier
  • Protino Ni-TED 150 packed columns (10)
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : imidazole, monosodium phosphate