Learn More
Macherey-Nagel™ Protino GST/4B Column Adaptor Sets
Click to view available options
Type:
1/4-28 Adapter Set
Inlet Adapter Set
Inlet Luer Adapter
Luer Adapter Set
M6 Adapter Set
For Use With (Equipment):
FPLC System, First Generation (Pharmacia)
MPLC System
Peristaltic pump
Syringe
Unit Size:
1 set
Each
Description
Protino™ GST/4B Columns
- Ready-to-use FPLC columns filled with Protino™ Glutathione Agarose 4B
- Best choice for cost-effective GST-tag protein purification
- Highest performance equivalent to Glutathione Sepharose™ 4B / GSTrap™ 4B columns
- Simply replace your current products, no optimization or protocol change necessary
- Suitable for small proteins, large protein complexes, proteins with low expression rates – universal use
- Market-leading performance for an unbeatable price
Specifications
Specifications
| Kit Contents | 1 x M6 female to 10-32 male; 1 x 10-32 female to M6 male |
| For Use With (Equipment) | FPLC System, First Generation (Pharmacia) |
| Quantity | 1 Set |
| For Use With (Application) | Purification of GST-tagged Proteins |
| Type | M6 Adapter Set |
| Column Type | Column Accessory |