Learn More
Macherey-Nagel™ Protino GST/4B Column Adaptor Sets
Product Code. 15860988
Click to view available options
Type:
1/4-28 Adapter Set
Inlet Adapter Set
Inlet Luer Adapter
Luer Adapter Set
M6 Adapter Set
For Use With (Equipment):
FPLC System, First Generation (Pharmacia)
MPLC System
Peristaltic pump
Syringe
Unit Size:
1 set
Each
Product Code. 15860988
Brand: Macherey-Nagel™ 745260
Only null left
Add to basket
Add to basket
Adapters for connecting Protino™ GST/4B columns to a system of choice (e.g. liquid chromatography system, peristaltic pump, syringe) for purification of GST-tagged proteins.
Protino™ GST/4B Columns
  • Ready-to-use FPLC columns filled with Protino™ Glutathione Agarose 4B
  • Best choice for cost-effective GST-tag protein purification
  • Highest performance equivalent to Glutathione Sepharose™ 4B / GSTrap™ 4B columns
  • Simply replace your current products, no optimization or protocol change necessary
  • Suitable for small proteins, large protein complexes, proteins with low expression rates – universal use
  • Market-leading performance for an unbeatable price
Specifications
Kit Contents 1 x M6 female to 10-32 male; 1 x 10-32 female to M6 male
For Use With (Equipment) FPLC System, First Generation (Pharmacia)
Quantity 1 Set
For Use With (Application) Purification of GST-tagged Proteins
Type M6 Adapter Set
Column Type Column Accessory