Learn More
R&D Systems™ Proteome Profiler Human Protease Array Kit
GREENER_CHOICE
Product Code. 18027631
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18027631 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18027631 Supplier R&D Systems™ Supplier No. ARY021B
  • ONLINE EXCLUSIVE PRICE
    732.00€
    691.95€ / 1 sets
    Save 40.05€
    5% Off
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
The Proteome Profiler Human Protease Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Human Protease Array Kit contains 4 membranes - each spotted in duplicate with 35 different protease antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit
Kit Contents 4 Array Membranes, 4-Well Multi-Dish, Array Buffers, Wash Buffer, Detection Antibody Cocktail, Streptavidin-HRP, Chemiluminescent Detection Reagents, Transparency Overlay Template, Detailed Protocol